Learn More
QED ENVIRONMENTAL SYSTEMS G110 Infrared CO2 Monitor includes Pump
Catalog No. NC1785663
Encompass_Preferred
The G110 infrared portable CO2 gas analyzer has been designed to monitor carbon dioxide for multiple applications including the brewing industry.
Catalog No. NC1785663 Supplier QED ENVIRONMENTAL SYSTEMS Supplier No. G11000L
The G110 infrared portable CO2 gas analyzer has been designed to monitor carbon dioxide for multiple applications including the brewing industry.