Learn More
World Precision Instrument CO2 Analyzer for Small Animals
SDP
Catalog No. 502538876
Encompass
The MICROCAPSTAR End-Tidal Carbon Dioxide Analyzer provides accurate end-tidal or continuous measurement of expired CO2 in animals as small as mice. It features very low sample flow requirements, rapid response time, and long-term stability. Respiratory rate (RR) is computed using the excursions of the CO2 waveform. The CO2 and RR measurements, as well as a trend plot of the end-tidal values, are displayed on the graphics LCD screen.
Catalog No. 50-253-8876 Supplier World Precision Instrument Supplier No. CWMICROCAPSTAR
Add to Cart
Add to Cart
Request a Quote
The MICROCAPSTAR End-Tidal Carbon Dioxide Analyzer provides accurate end-tidal or continuous measurement of expired CO2 in animals as small as mice. It features very low sample flow requirements, rapid response time, and long-term stability. Respiratory rate (RR) is computed using the excursions of the CO2 waveform. The CO2 and RR measurements, as well as a trend plot of the end-tidal values, are displayed on the graphics LCD screen.