Learn More
Fisherbrand™ CO2 Backup Systems for Isotemp Chest Ultra-Low Temperature Freezers
Catalog No. p-7227920
Change view
Click to view available options
For Use With (Equipment):
Isotemp 13 and 20 cu. ft. Chest Ultra-Low Temperature Freezer
Isotemp 3 cu. ft. Chest Ultra-Low Temperature Freezer
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
332353G04 Isotemp 3 cu. ft. Chest Ultra-Low Temperature Freezer
332353G08 Isotemp 13 and 20 cu. ft. Chest Ultra-Low Temperature Freezer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 332353G04 Supplier Fisherbrand™ Supplier No. 332353G04
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Ensure optimal security for critical samples by using Fisherbrand™ Isotemp CO2 Backup Systems for Ultra-Low Temperature Freezers.
  • Maintains temperature down to -67°C with CO2
TRUSTED_SUSTAINABILITY
Specifications
Description Factory Installed
Temperature Range -67°C
For Use With (Equipment) Isotemp 3 cu. ft. Chest Ultra-Low Temperature Freezer