Learn More
PCE AMERICAS INC Portable CO2 Meter 0 … 9999 ppm
SDP
Catalog No. NC3654613
Encompass_Preferred
"CO2 analyser with data memory for 5000 measuring points / Temperature, humidity /Export of the saved measurement data in CSV format / Battery and mains operation /1/4 inch tripod mount / Measuring range (CO²) 0 ... 9999 ppm, (CO) 0 ... 1000 ppmThe CO2 analyser has a measuring range of 0 ... 9999 ppm. In addition to the carbon dioxide measurement, the air quality detector also has a carbon monoxide (CO) sensor with a measuring range of 0 ... 1000 ppm and a temperature and humidity sensor. The air quality detector already covers a large range of measuring tasks. The air quality detector is therefore used, for example, in occupational safety to check the climatic conditions in office workplaces."
Catalog No. NC3654613 Supplier PCE AMERICAS INC Supplier No. PCECOG 10
"CO2 analyser with data memory for 5000 measuring points / Temperature, humidity /Export of the saved measurement data in CSV format / Battery and mains operation /1/4 inch tripod mount / Measuring range (CO²) 0 ... 9999 ppm, (CO) 0 ... 1000 ppmThe CO2 analyser has a measuring range of 0 ... 9999 ppm. In addition to the carbon dioxide measurement, the air quality detector also has a carbon monoxide (CO) sensor with a measuring range of 0 ... 1000 ppm and a temperature and humidity sensor. The air quality detector already covers a large range of measuring tasks. The air quality detector is therefore used, for example, in occupational safety to check the climatic conditions in office workplaces."