Learn More
Bound Tree Medical CO2 DETECTOR EASY CAP EA
Catalog No. NC0079685
Encompass_Preferred
Easy Cap II or CO2 Detector is a single patient use to verify endotracheal tube placement unit. Responds to exhaled CO2 with a simple color change from purple to yellow. Breath-to-breath response.
Catalog No. NC0079685 Supplier Bound Tree Medical Supplier No. 530024
Easy Cap II or CO2 Detector is a single patient use to verify endotracheal tube placement unit. Responds to exhaled CO2 with a simple color change from purple to yellow. Breath-to-breath response.