Learn More
Thermo Scientific™ CO2 Gas Guard
Catalog No. 13998120 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
3130/3131, 3140/3141 incubators
Catalog No. For Use With (Equipment)
13-998-120 3130/3131, 3140/3141 incubators
1 options
Catalog No. 13-998-120 Supplier Thermo Scientific™ Supplier No. 190642
Add to Cart
Add to Cart
For Use With: 3130/3131, 3140/3141 Incubators
Minimize the need to frequently change out cylinders and reduce the risk of running out of gas. Thermo Scientific™ Gas Guards for CO2 and LN2 Tanks are factory installed and built in to specified CO2 incubators; specify when ordering the incubator. Fitting unobtrusively inside the incubator casing, these systems interface directly with unit power supply and control systems.
  • Factory-installed systems monitor CO2 or N2 pressure in source tanks
  • Automatically switch from one cylinder to another when supply in first tank is exhausted (pressure falls below 15psi)
  • Audible and visual alarms on incubator control panel alert to tank switch-overs and to when both tanks are low
TRUSTED_SUSTAINABILITY
Specifications
Description Gas Guard, CO2
For Use With (Equipment) 3130/3131, 3140/3141 incubators
Lighting LED
Type CO2 Gas Guard