Learn More
Sheldon Manufacturing Inc Co2 Dry Inc, 31 cuft, 115v
SDP
Catalog No. NC1642410
Encompass_Preferred
Model SCO31 large capacity dry CO2 incubator is ideal for roller bottle apparatus and high-volume tissue culture applications and cell harvesting. It provides a precisely controlled, accurate, pure environment throughout the culturing process. Also available in a 40 cu.ft./1125 L size (Model SCO40) and 58 cu.ft./1641 L size (Model SCO58). Includes a two-year limited warranty. Standard Design Features: Infrared (IR) CO2_Sensor is extremely quick to recover CO2 levels after door openings and is not affected by temperature or humidity levels, Unique air jacket design provides excellent temperature uniformity of +/-0.5°C at 37°C, Ideal for roller bottle apparatus includes a flip-out ramp, Corrosion resistant, easily cleanable stainless steel chamber, Viewing window in door, 1 interior electrical outlet
Catalog No. NC1642410 Supplier Sheldon Manufacturing Inc Supplier No. SCO31
Add to Cart
Add to Cart
Request a Quote
Model SCO31 large capacity dry CO2 incubator is ideal for roller bottle apparatus and high-volume tissue culture applications and cell harvesting. It provides a precisely controlled, accurate, pure environment throughout the culturing process. Also available in a 40 cu.ft./1125 L size (Model SCO40) and 58 cu.ft./1641 L size (Model SCO58). Includes a two-year limited warranty. Standard Design Features: Infrared (IR) CO2_Sensor is extremely quick to recover CO2 levels after door openings and is not affected by temperature or humidity levels, Unique air jacket design provides excellent temperature uniformity of +/-0.5°C at 37°C, Ideal for roller bottle apparatus includes a flip-out ramp, Corrosion resistant, easily cleanable stainless steel chamber, Viewing window in door, 1 interior electrical outlet