Learn More
PHCbi CO2 Incubator
Catalog No. 502720068
Encompass_Preferred
PHCbi brand 8.1 cu.ft (230 L) Cell-IQTM series CO2 incubator. Utilizes direct heat and air jacket syste, IR2 sensor, SafeCellTM UV and InCu-saFe, copper-infused stainless stell interior, and offers an optional high speed H2O2 decontamination system.Temperature Range+5°C above ambient to +50°C
Catalog No. 50-272-0068 Supplier PHCbi Supplier No. MCO230AICUVLGPA
Add to Cart
Add to Cart
Request a Quote
PHCbi brand 8.1 cu.ft (230 L) Cell-IQTM series CO2 incubator. Utilizes direct heat and air jacket syste, IR2 sensor, SafeCellTM UV and InCu-saFe, copper-infused stainless stell interior, and offers an optional high speed H2O2 decontamination system.Temperature Range+5°C above ambient to +50°C