Learn More
PHCbi CO2 Incubator
Supplier:  PHCbi MCO170ACLPA
Encompass_Preferred
PHCbi brand 5.8 cu.ft (165L) CytoGrow CO2 incubator. Desgined with standard, passive contamination control via the patented inCu-saFe (R) cooper infused stainless stell interior, this stackable unit offers a removable shelf channel system which allows for easy cleaning, eliminating the need for shelf brackets and clips. Temperature Range+5°C above ambient to +50°C
Catalog No. 50-272-0071
Add to Cart
Request a Quote