Learn More
RWD LIFE SCIENCE INC CO2 incubator D181, volume 188L; 140℃ setrilization; with humidity monitoring
SDP
Catalog No. NC3404383
Encompass_Preferred
D181 CO2 incubator provides a more suitable culture environment for cells by controlling temperature, CO2 concentration precisely and by maintaining a high relative humidity (rH). It is an indispensable cell culture tool in the fields of stem cell research, immunology research, neurobiology research and cancer research.
Catalog No. NC3404383 Supplier RWD LIFE SCIENCE INC Supplier No. D181
Add to Cart
Add to Cart
Request a Quote
D181 CO2 incubator provides a more suitable culture environment for cells by controlling temperature, CO2 concentration precisely and by maintaining a high relative humidity (rH). It is an indispensable cell culture tool in the fields of stem cell research, immunology research, neurobiology research and cancer research.