Learn More
Nuaire Inc CO2 Incubator Tubing Kit
SDP
Catalog No. NC1684581
Encompass
Connect your CO2 or N2 gas tanks to your NuAire CO2 Incubator with this tubing kit featuring 6-feet in total tubing and inline filtration.
Catalog No. NC1684581 Supplier Nuaire Inc Supplier No. NU1556
Connect your CO2 or N2 gas tanks to your NuAire CO2 Incubator with this tubing kit featuring 6-feet in total tubing and inline filtration.