Learn More
PCE AMERICAS INC CO2 Meter measuring range up to 9999 ppm
SDP
Catalog No. NC3654611
Encompass_Preferred
"hermometer for tabletop operation / Measures temperature, humidity and CO2 content of the air /CO2 measuring range from 0… 9999 ppm / Traffic light function / Touchscreen / USB supplyWith the CO2 meter you can ensure the right climate in your office. In addition to the CO2 content of the air, the CO2 meter measures the relative humidity and room temperature. The CO2 meter shows carbon dioxide concentrations up to 9999 ppm, temperatures from -10… 50 °C, and relative humidity values ​​from 20 … 95%. On the 3.5"" display of the CO2 meter, the corresponding values can be read off directly, and the color of the CO2 display changes to provide an objective statement about the quality of the room air based on the CO2 content. This makes it particularly easy to assess the indoor air quality using the CO2 meter. For these reasons, the CO2 meter is particularly suitable for offices, schools, public buildings or hospitals, etc.
Catalog No. NC3654611 Supplier PCE AMERICAS INC Supplier No. PCECMM 8
"hermometer for tabletop operation / Measures temperature, humidity and CO2 content of the air /CO2 measuring range from 0… 9999 ppm / Traffic light function / Touchscreen / USB supplyWith the CO2 meter you can ensure the right climate in your office. In addition to the CO2 content of the air, the CO2 meter measures the relative humidity and room temperature. The CO2 meter shows carbon dioxide concentrations up to 9999 ppm, temperatures from -10… 50 °C, and relative humidity values ​​from 20 … 95%. On the 3.5"" display of the CO2 meter, the corresponding values can be read off directly, and the color of the CO2 display changes to provide an objective statement about the quality of the room air based on the CO2 content. This makes it particularly easy to assess the indoor air quality using the CO2 meter. For these reasons, the CO2 meter is particularly suitable for offices, schools, public buildings or hospitals, etc.