Learn More
PCE AMERICAS INC CO2 Meter measuring range up to 5000 ppm
SDP
Catalog No. NC3654609
Encompass_Preferred
The CO2 content in the air is an important measure for checking the ventilation behavior in offices, classrooms, lecture halls, etc. The CO2 / carbon dioxide measuring device helps to ensure that, for example, there is regular ventilation for employees in the office. The CO2 / carbon dioxide measuring device checks indoor air quality via the CO2 content. If the CO2 concentration is below 1000 ppm, the indoor air quality is optimal. Good indoor air quality helps to promote concentration. It is also ensured that the aerosol concentration in the air is low, as the aerosols are repeatedly mixed with fresh air. The CO2 / carbon dioxide measuring device shows the air quality as a numerical measured value and as a traffic light indicator. At the push of a button, the CO2 content of the last five hours can be shown on the display. In addition to the CO2 measuring function, the CO2 / carbon dioxide measuring device is equipped with a sensor.
Catalog No. NC3654609 Supplier PCE AMERICAS INC Supplier No. PCECMM 5
The CO2 content in the air is an important measure for checking the ventilation behavior in offices, classrooms, lecture halls, etc. The CO2 / carbon dioxide measuring device helps to ensure that, for example, there is regular ventilation for employees in the office. The CO2 / carbon dioxide measuring device checks indoor air quality via the CO2 content. If the CO2 concentration is below 1000 ppm, the indoor air quality is optimal. Good indoor air quality helps to promote concentration. It is also ensured that the aerosol concentration in the air is low, as the aerosols are repeatedly mixed with fresh air. The CO2 / carbon dioxide measuring device shows the air quality as a numerical measured value and as a traffic light indicator. At the push of a button, the CO2 content of the last five hours can be shown on the display. In addition to the CO2 measuring function, the CO2 / carbon dioxide measuring device is equipped with a sensor.