Learn More
Cole-Parmer Data Logging Carbon Dioxide Data Logging Carbon Dioxide Monitor with Temperature and Humiditywith Temperature and Humidity
Catalog No. NC3306575 Shop All Cole-Parmer Products
Encompass_Preferred
Logs up to 15,999 points of dataLarge display shows carbon dioxide level, temperature, humidity, date, and time15 degree tilt angle is easy to see and readProvides an indoor air quality indicationStable NDIR sensor for carbon dioxide detectionVisible and audible carbon dioxide alarm1 year warranty
Catalog No. NC3306575 Supplier Cole-Parmer Supplier No. 9456000
Logs up to 15,999 points of dataLarge display shows carbon dioxide level, temperature, humidity, date, and time15 degree tilt angle is easy to see and readProvides an indoor air quality indicationStable NDIR sensor for carbon dioxide detectionVisible and audible carbon dioxide alarm1 year warranty