Learn More
Euthanex The EP-PS50 Tamper-Proof Preset CO2 Regulator with 0.5 to 12 LPM Range Flow Meter
Catalog No. NC0479330
Encompass
The EP-PS50 Tamper-Proof Preset Regulator incorporates a flow meter and connects directly to the CO2 cylinder. Factory preset to your specific cage size to ensure AVMA compliance. The user simply needs to turn a valve on or off to administer gas at the facility"s approved flow rate. Visual indicator ensure corrects gas flow rates. Flow rate range is 0.5 to 12 LPM. Designated on-site personnel may change the flow rate if guidelines or facility SOP's change. Connects directly to CO2 cylinder with CGA-320 fitting (optional CGA-940 Yoke fitting available). Includes 9 foot hose with quick disconnect fitting.
Catalog No. NC0479330 Supplier Euthanex Supplier No. EPPS50
The EP-PS50 Tamper-Proof Preset Regulator incorporates a flow meter and connects directly to the CO2 cylinder. Factory preset to your specific cage size to ensure AVMA compliance. The user simply needs to turn a valve on or off to administer gas at the facility"s approved flow rate. Visual indicator ensure corrects gas flow rates. Flow rate range is 0.5 to 12 LPM. Designated on-site personnel may change the flow rate if guidelines or facility SOP's change. Connects directly to CO2 cylinder with CGA-320 fitting (optional CGA-940 Yoke fitting available). Includes 9 foot hose with quick disconnect fitting.