Learn More
Medchemexpress LLC CO23 | 1370363-74-8 | 98.0% | 592.17 g/mol | C19H18I2N2O4 | 5 MG
SDP
Catalog No. 5000262824
Encompass_Preferred
CO23 is a selective thyroid hormone receptor alpha (TRα) agonist used in research to study growth, development, and thyroid signaling. It has been reported to cross the blood-brain barrier and is supplied as a high-purity research chemical for in vitro and in vivo studies.
  • Selective TRα agonist for targeted thyroid receptor modulation.
  • Demonstrated blood-brain barrier penetration.
  • High purity (98.0%) for reliable experimental results.
  • Soluble in DMSO at 250 mg/mL; ultrasonic assistance recommended.
  • Solid, off-white to light yellow appearance; powder stable at -20 °C.
Catalog No. 50-002-62824 Supplier Medchemexpress LLC Supplier No. HY1300125MG
CO23 is a selective thyroid hormone receptor alpha (TRα) agonist used in research to study growth, development, and thyroid signaling. It has been reported to cross the blood-brain barrier and is supplied as a high-purity research chemical for in vitro and in vivo studies.
  • Selective TRα agonist for targeted thyroid receptor modulation.
  • Demonstrated blood-brain barrier penetration.
  • High purity (98.0%) for reliable experimental results.
  • Soluble in DMSO at 250 mg/mL; ultrasonic assistance recommended.
  • Solid, off-white to light yellow appearance; powder stable at -20 °C.