Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Co23 | 1370363-74-8 | 98.0% | C19H18I2N2O4 | 10MG
SDP

Catalog No. 5000260533
Encompass_Preferred

CO23 is a selective thyroid hormone receptor (TR) α agonist for research applications. It is an off-white to light-yellow solid with reported ability to cross the blood-brain barrier and is used to study growth and developmental regulation mediated by TRα. The compound is provided with defined purity and storage recommendations for stable handling.

  • Selective thyroid hormone receptor (TRα) agonist.
  • Reported to cross the blood-brain barrier.
  • Off-white to light-yellow solid appearance.
  • Purity 98.0% as supplied.
  • Available as solid and as a 10 mM solution in DMSO.
  • Storage recommended at -20°C for powder and -80°C in solvent for long term.

Catalog No. 50-002-60533 Supplier Medchemexpress LLC Supplier No. HY13001210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CO23 is a selective thyroid hormone receptor (TR) α agonist for research applications. It is an off-white to light-yellow solid with reported ability to cross the blood-brain barrier and is used to study growth and developmental regulation mediated by TRα. The compound is provided with defined purity and storage recommendations for stable handling.

  • Selective thyroid hormone receptor (TRα) agonist.
  • Reported to cross the blood-brain barrier.
  • Off-white to light-yellow solid appearance.
  • Purity 98.0% as supplied.
  • Available as solid and as a 10 mM solution in DMSO.
  • Storage recommended at -20°C for powder and -80°C in solvent for long term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.