Learn More
Pacific Hemostasis™ Coagulation Control Level 2 (Abnormal), 10 x 1mL
Provide accurate, reproducible confirmation of coagulation test system performance with these coagulation controls.
Catalog No. 17-621-4
This product is regulated by the federal government, and may not be ordered online.This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.
This item is not returnable. View return policy
Description
Description
Provide accurate, reproducible confirmation of coagulation test system performance with Thermo Scientific™ Pacific Hemostasis™ Coagulation Controls. Controls can be run on most coagulation systems and are formulated to reflect expected patient results.
Coagulation and Heparin control plasmas are prepared from fresh pools of human plasma.
Coagulation Control Plasmas:
  • Normal control (Level 1) produces normal PT and aPTT times
  • Abnormal controls monitor therapeutic (Level 2) and highly elevated (Level 3) PT/aPTT times 
Heparin Control Plasmas:
  • Simulate low (Level 1) and elevated (Level 2) plasma levels of Heparin
  • Use to monitor and evaluate patients on Heparin therapy
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions