Learn More
ACROBiosystems Cynomolgus Coagulation factor IX / F9 Protein, His Tag (active enzyme, MALS verified)
Supplier:  ACROBiosystems FIXC52H9100UG
Encompass_Preferred
Cynomolgus Coagulation factor IX, His Tag (FIX-C52H9) is expressed from human 293 cells (HEK293). It contains AA Thr 29 - Thr 461 (Accession # G7Q1U2-1).
Catalog No. 50-233-1142