Learn More
Sino Biological Coagulation Factor IX / FIX / F9 Antibody, Rabbit PAb 100uL
Supplier:  Sino Biological 11503RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Coagulation Factor IX / FIX / F9 (rh F9; Catalog#11503-H08H; AAB59620.1; Met1-Thr461). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-224-8024