Learn More
Cusabio Technology LLC coagulation factor VFV
Catalog No. 5000051105
Encompass_Preferred
This kit detects F5 (Human coagulation factor ) in serum plasma and tissue homogenates using a quantitative sandwich ELISA method It offers a detection range of 3 12 ng/mL to 200 ng/mL with a sensitivity of 0 78 ng/mL enabling precise measurement of F5 concentrations in biological samples The assay requires a sample volume of 50-100 L and can be completed within 1-5 hours Detection is performed at 450 nm ensuring reliable and specific results for research or diagnostic applications Designed for flexibility this kit provides a standardized approach to analyze coagulation factor levels across multiple sample types while maintaining high reproducibility and accuracy
Catalog No. 50-000-51105 Supplier Cusabio Technology LLC Supplier No. CSBE14302H
This kit detects F5 (Human coagulation factor ) in serum plasma and tissue homogenates using a quantitative sandwich ELISA method It offers a detection range of 3 12 ng/mL to 200 ng/mL with a sensitivity of 0 78 ng/mL enabling precise measurement of F5 concentrations in biological samples The assay requires a sample volume of 50-100 L and can be completed within 1-5 hours Detection is performed at 450 nm ensuring reliable and specific results for research or diagnostic applications Designed for flexibility this kit provides a standardized approach to analyze coagulation factor levels across multiple sample types while maintaining high reproducibility and accuracy