Learn More
Cusabio Technology LLC Coagulation factor VIIF7
Catalog No. 5000051142
Encompass_Preferred
This kit detects F7 in serum plasma and tissue homogenates with a detection range of 0 78 ng/mL to 50 ng/mL and a sensitivity of 0 195 ng/mL It employs a quantitative sandwich ELISA method providing reliable results within 1 to 5 hours The assay requires 50-100 L of sample per test and measures absorbance at 450 nm Designed for accurate measurement of Rat Coagulation Factor VII levels it is suitable for researchers studying coagulation factors or related pathways The optimized protocol ensures consistent performance across diverse biological samples while maintaining high specificity and reproducibility
Catalog No. 50-000-51142 Supplier Cusabio Technology LLC Supplier No. CSBEL007930RA
This kit detects F7 in serum plasma and tissue homogenates with a detection range of 0 78 ng/mL to 50 ng/mL and a sensitivity of 0 195 ng/mL It employs a quantitative sandwich ELISA method providing reliable results within 1 to 5 hours The assay requires 50-100 L of sample per test and measures absorbance at 450 nm Designed for accurate measurement of Rat Coagulation Factor VII levels it is suitable for researchers studying coagulation factors or related pathways The optimized protocol ensures consistent performance across diverse biological samples while maintaining high specificity and reproducibility