Learn More
Cayman Chemical Coagulation Factor IX (human, recombinant), Purity: ≥95% as determined by SDS-PAGE, 1 mg
SDP
Catalog No. 502950423
Encompass_Preferred
Source: Active recombinant human C-terminal His-tagged coagulation factor IX expressed in HEK293 cells • Amino acids: 29-461 • MW: 50 kDa
Catalog No. 50-295-0423 Supplier Cayman Chemical Supplier No. 318381 MG
Source: Active recombinant human C-terminal His-tagged coagulation factor IX expressed in HEK293 cells • Amino acids: 29-461 • MW: 50 kDa