Learn More
Cayman Chemical CoagulatIon Factr IXhumn50ug
SDP
Catalog No. 502950469
Encompass_Preferred
Source: Active recombinant human C-terminal His-tagged coagulation factor IX expressed in HEK293 cells • Amino acids: 29-461 • MW: 50 kDa
Catalog No. 50-295-0469 Supplier Cayman Chemical Supplier No. 3183850 UG
Source: Active recombinant human C-terminal His-tagged coagulation factor IX expressed in HEK293 cells • Amino acids: 29-461 • MW: 50 kDa