Learn More
Cayman Chemical CoagulatIon Factr Xhumn500ug
SDP
Catalog No. 502942152
Encompass_Preferred
Source: Recombinant human C-terminal His-tagged coagulation factor X expressed in insect cells • Amino acids: 1-488 (full length) • MW: 52.8 kDa
Catalog No. 50-294-2152 Supplier Cayman Chemical Supplier No. 40252500 UG
Source: Recombinant human C-terminal His-tagged coagulation factor X expressed in insect cells • Amino acids: 1-488 (full length) • MW: 52.8 kDa