Learn More
Diapharma Group, Inc. Chromogenix Coamatic Factor VIII, 120 tests
SDP
Catalog No. NC9582546
Encompass
Chromogenix Coamatic Factor VIII is a chromogenic assay kit for the photometric determination of factor VIII activity in plasma, such as when identifying factor VIII deficiency or monitoring patients on replacement therapy as well as for potency estimation of FVIII concentrates. This Factor VIII test kit fulfills the requirements of the European Pharmacopoeia for factor VIII concentrate testing. Co-lyophilization of reagents for convenient handling. Reliable tool for hemophilia classification. No interference from heparin levels up to 1.5 IU/ml. The Factor VIII kit has two measuring ranges.
Catalog No. NC9582546 Supplier Diapharma Group, Inc. Supplier No. K822585
Chromogenix Coamatic Factor VIII is a chromogenic assay kit for the photometric determination of factor VIII activity in plasma, such as when identifying factor VIII deficiency or monitoring patients on replacement therapy as well as for potency estimation of FVIII concentrates. This Factor VIII test kit fulfills the requirements of the European Pharmacopoeia for factor VIII concentrate testing. Co-lyophilization of reagents for convenient handling. Reliable tool for hemophilia classification. No interference from heparin levels up to 1.5 IU/ml. The Factor VIII kit has two measuring ranges.