Learn More
MSC Coast Handheld Flashlight, 640 Lumens, Twist Focus, Aluminum, 1 ea
Catalog No. 5000795583
Encompass
The Coast PX1 flashlight features PURE BEAM® optics with TWIST FOCUS™, allowing seamless transitions between a wide flood beam and a long-range spot beam. This versatile flashlight provides up to 640 lumens of light output and is designed for reliable performance in various conditions.
  • Pure Beam Optic for seamless flood to spot beam transitions
  • Twist Focus to easily adjust beam shape
  • IP54 rated for water and dust resistance
  • Dual power capability using AAA batteries or optional rechargeable battery
  • Offers up to 640 lumens of light
  • Maximum run time of 2.75 hours on high setting
  • Constructed with a durable aluminum body
Catalog No. 50-007-95583 Supplier MSC Supplier No. 35058890
The Coast PX1 flashlight features PURE BEAM® optics with TWIST FOCUS™, allowing seamless transitions between a wide flood beam and a long-range spot beam. This versatile flashlight provides up to 640 lumens of light output and is designed for reliable performance in various conditions.
  • Pure Beam Optic for seamless flood to spot beam transitions
  • Twist Focus to easily adjust beam shape
  • IP54 rated for water and dust resistance
  • Dual power capability using AAA batteries or optional rechargeable battery
  • Offers up to 640 lumens of light
  • Maximum run time of 2.75 hours on high setting
  • Constructed with a durable aluminum body