Learn More
Medchemexpress LLC COASY Human His 1mg | 1mg
SDP
Catalog No. 5000186034
Encompass_Preferred
COASY protein is a bifunctional enzyme that plays a key role in catalyzing the fourth and fifth steps of the CoA biosynthetic pathway The coaD domain promotes the phosphopantothenate adenylyltransferase activity of the fourth reaction and the coaE domain drives the dephosphoCoA kinase activity of the fifth reaction COASY Protein Human (His) is the recombinant human-derived COASY protein expressed by E coli with N-His labeled tag
Catalog No. 50-001-86034 Supplier Medchemexpress LLC Supplier No. HYP768431MG
COASY protein is a bifunctional enzyme that plays a key role in catalyzing the fourth and fifth steps of the CoA biosynthetic pathway The coaD domain promotes the phosphopantothenate adenylyltransferase activity of the fourth reaction and the coaE domain drives the dephosphoCoA kinase activity of the fifth reaction COASY Protein Human (His) is the recombinant human-derived COASY protein expressed by E coli with N-His labeled tag