Learn More
Grainger Coat No Insulation White M
Catalog No. 502753636
Encompass_Preferred
The Grainger 14H701 is a 14-inch diameter, 7-blade circular saw blade with a 1-inch arbor hole and a 0.125-inch kerf width. It has a hook angle of 7 degrees and is made from high-speed steel with a 0.042-inch body thickness. This general purpose blade is designed for cross-cutting and ripping of wood, plywood, and non-ferrous metals. It has a maximum speed of 6,300 RPM. The Grainger 14H701 circular saw blade is ideal for use with portable circular saws, radial arm saws, and table saws.
Catalog No. 50-275-3636 Supplier Grainger Supplier No. 14H701
The Grainger 14H701 is a 14-inch diameter, 7-blade circular saw blade with a 1-inch arbor hole and a 0.125-inch kerf width. It has a hook angle of 7 degrees and is made from high-speed steel with a 0.042-inch body thickness. This general purpose blade is designed for cross-cutting and ripping of wood, plywood, and non-ferrous metals. It has a maximum speed of 6,300 RPM. The Grainger 14H701 circular saw blade is ideal for use with portable circular saws, radial arm saws, and table saws.