Learn More
Grainger Coat No Insulation White S
Catalog No. 502748867
Encompass_Preferred
The Grainger 14H708 is a heavy duty industrial grade bench vise that features a 6 inch jaw width capacity, 4 inch jaw opening capacity, and a 360 degree swivel base. It has an anvil working surface and is constructed from high strength cast iron with a powder coat finish. This vise can securely clamp a variety of workpieces up to 6 inches wide and 4 inches tall. With its sturdy build and swiveling base, the 14H708 bench vise is well suited for metalworking, assembly, and other industrial applications.
Catalog No. 50-274-8867 Supplier Grainger Supplier No. 14H708
The Grainger 14H708 is a heavy duty industrial grade bench vise that features a 6 inch jaw width capacity, 4 inch jaw opening capacity, and a 360 degree swivel base. It has an anvil working surface and is constructed from high strength cast iron with a powder coat finish. This vise can securely clamp a variety of workpieces up to 6 inches wide and 4 inches tall. With its sturdy build and swiveling base, the 14H708 bench vise is well suited for metalworking, assembly, and other industrial applications.