Learn More
A-M Systems COATD PLTNM-IRIDIUM WIRE, 10FT
SDP
Catalog No. NC2263017
Encompass_Preferred
Coated Platinum-Iridium Wire.This wire features a 90% Platinum and 10% Iridium composition and is widely used in the assembly of microelectrode probes. Iridium, a Platinum group metal, is very dense and hard, imparting added strength to the wire. 10 feet (3 m) per spool.
This item is not returnable. View return policy
Catalog No. NC2263017 Supplier A-M Systems Supplier No. 776000
Coated Platinum-Iridium Wire.This wire features a 90% Platinum and 10% Iridium composition and is widely used in the assembly of microelectrode probes. Iridium, a Platinum group metal, is very dense and hard, imparting added strength to the wire. 10 feet (3 m) per spool.