Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Coated GC Packing, Phase 0.3 Carbowax 20M + 0.1 H3PO4, Matrix 60/80 Carbopack C Support

Catalog No. 111003575 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
15 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-3575 15 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-3575 Supplier MilliporeSigma™ Supelco™ Supplier No. 11825U

missing translation for 'validMainframeMsg'

Ready-to-use coated packing for those wanting to pack their own columns.

The choice of coating material is based on the stationary phase of the GC.
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottle
Quantity 15 g
For Use With (Application) Gas Chromatography (GC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.