Learn More
MilliporeSigma™ Supelco™ Coated GC Packing, Phase 1 SP™-1510, Matrix 60/80 Carbopack B Support
Catalog No. 111003612 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
15 g
Catalog No. Quantity
11-100-3612 15 g
1 options
Catalog No. 11-100-3612 Supplier MilliporeSigma™ Supelco™ Supplier No. 11809
Add to Cart
Add to Cart
Ready-to-use coated packing for those wanting to pack their own columns.
The choice of coating material is based on the stationary phase of the GC.
TRUSTED_SUSTAINABILITY
Specifications
Packaging Bottle
Quantity 15 g
For Use With (Application) Gas Chromatography (GC)
SP is a registered trademark of Sigma-Aldrich Co. LLC