Learn More
MilliporeSigma™ Supelco™ Coated GC Packing, Phase 5 Carbowax 20M, Matrix 60/80 Carbopack B Support
Catalog No. 111003611 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
15 g
Catalog No. Quantity
11-100-3611 15 g
1 options
Catalog No. 11-100-3611 Supplier MilliporeSigma™ Supelco™ Supplier No. 11766
Add to Cart
Add to Cart
Ready-to-use coated packing for those wanting to pack their own columns.
The choice of coating material is based on the stationary phase of the GC.
TRUSTED_SUSTAINABILITY
Specifications
Packaging Bottle
Quantity 15 g
For Use With (Application) Gas Chromatography (GC)