Learn More
Delta Technologies Ltd Polished Float Glass, 25 x 75 x 1.1 mm, SiO2 passivated/Indium Tin Oxide coated on polished surface, Rs = 30 - 60 ohms, cut edges. Pack of 25 slides.
SDP
Catalog No. NC1662759
Encompass
ITO Coated Slides, Polished Float Glass, 25 x 75 x 1.1 mm, SiO2 passivated/Indium Tin Oxide coated on polished surface, resistance: 30-60 ohms/sq. This is for a pack of 25 slides. Delta Technologies, Limited offers a variety of transparent, conductive indium tin oxide (ITO), flourine-doped tin oxide (FTO) coated, and uncoated products on several different substrates, custom cut to any dimension required.
Catalog No. NC1662759 Supplier Delta Technologies Ltd Supplier No. CG81INS11525
ITO Coated Slides, Polished Float Glass, 25 x 75 x 1.1 mm, SiO2 passivated/Indium Tin Oxide coated on polished surface, resistance: 30-60 ohms/sq. This is for a pack of 25 slides. Delta Technologies, Limited offers a variety of transparent, conductive indium tin oxide (ITO), flourine-doped tin oxide (FTO) coated, and uncoated products on several different substrates, custom cut to any dimension required.