Learn More
Diapharma Group, Inc. Chromogenix Coatest Heparin, 400 tests
SDP
Catalog No. NC9417941
Encompass
Chromogenix Coatest Heparin is a chromogenic assay kit for the in vtiro diagnostic photometric determination of heparin in human plasma. The Coatest Heparin test kit determines the anti-FXa activity of LMW heparin and UF heparin. Excellent reagent stability. Suitable for both large and small laboratories.
Catalog No. NC9417941 Supplier Diapharma Group, Inc. Supplier No. K255539
Chromogenix Coatest Heparin is a chromogenic assay kit for the in vtiro diagnostic photometric determination of heparin in human plasma. The Coatest Heparin test kit determines the anti-FXa activity of LMW heparin and UF heparin. Excellent reagent stability. Suitable for both large and small laboratories.