Learn More
Seracare Life Sciences Inc COATING SOLUTION CONCENTRATE
Catalog No. 5067441
Encompass_Preferred
Convenient 10X buffer concentrate used to maximize binding to the solid phase in microwell ELISA. Formulated to provide optimal pH and ionic strength for binding of antibodies and some antigens. Compatible with most assay systems.
This item is not returnable. View return policy
Catalog No. 50-674-41 Supplier Seracare Life Sciences Inc Supplier No. 51500014
Convenient 10X buffer concentrate used to maximize binding to the solid phase in microwell ELISA. Formulated to provide optimal pH and ionic strength for binding of antibodies and some antigens. Compatible with most assay systems.