Learn More
Grainger Coaxial Cable RG-6 10 ft Black
Catalog No. 502749131
Encompass_Preferred
The Monoprice 18 AWG RG6 Coaxial Cable is a high quality coaxial cable for connecting video devices. It has an 18 AWG center conductor made of bare copper clad steel, surrounded by polyethylene dielectric insulation. The cable has a foil shield and braided shield for maximum signal protection from EMI and RFI interference. It has an RG6 type connector and is CL2 rated for in-wall installation. The cable is available in multiple lengths up to 500 ft. Overall this coaxial cable delivers clear, high resolution video signal transmission.
Catalog No. 50-274-9131 Supplier Grainger Supplier No. 14C393
The Monoprice 18 AWG RG6 Coaxial Cable is a high quality coaxial cable for connecting video devices. It has an 18 AWG center conductor made of bare copper clad steel, surrounded by polyethylene dielectric insulation. The cable has a foil shield and braided shield for maximum signal protection from EMI and RFI interference. It has an RG6 type connector and is CL2 rated for in-wall installation. The cable is available in multiple lengths up to 500 ft. Overall this coaxial cable delivers clear, high resolution video signal transmission.