Learn More
Grainger Coaxial Cable RG-6 15 ft Black
Catalog No. 502753227
Encompass_Preferred
The Monoprice 18 AWG Coaxial Cable is a high quality coaxial cable for connecting audio/video devices. It has an 18 AWG copper clad steel center conductor, polyethylene dielectric insulation, aluminum foil shielding with 65% tinned copper braid shielding, and a PVC jacket. This cable is CL2 rated for in-wall installation. It has low signal loss and supports digital HD video resolutions up to 1080p as well as all analog video formats. The cable has gold plated F-type connectors and is available in lengths from 1 to 100 feet. It's designed for connecting devices like satellite receivers, antennas, DVRs, TVs, and more.
Catalog No. 50-275-3227 Supplier Grainger Supplier No. 14C395
The Monoprice 18 AWG Coaxial Cable is a high quality coaxial cable for connecting audio/video devices. It has an 18 AWG copper clad steel center conductor, polyethylene dielectric insulation, aluminum foil shielding with 65% tinned copper braid shielding, and a PVC jacket. This cable is CL2 rated for in-wall installation. It has low signal loss and supports digital HD video resolutions up to 1080p as well as all analog video formats. The cable has gold plated F-type connectors and is available in lengths from 1 to 100 feet. It's designed for connecting devices like satellite receivers, antennas, DVRs, TVs, and more.