Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SI Analytics™ Coaxial Cables

Catalog No. 15177899 Shop All Yellow Springs Instrument (YSI) Products
Change view
Click to view available options
Length (Metric) Cable:
1 m
2 m
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Cable
15-177-899 2 m
15-177-898 1 m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 15-177-899 Supplier SI Analytics™ Supplier No. 285122464
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For Plug Head Electrodes

TRUSTED_SUSTAINABILITY

Specifications

Connector Types DIN Instrument Plug (A)
Type Electrode Cable (L)
Length (Metric) Cable 2 m
Model L 2 A
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.