Learn More
Norell Inc Coaxial Insert - 4mm TUBE W/50mm STEM. Includes one S-5-400-7 NMR tube to be used with the Coaxial Insert. 50mm Stem OD is 2mm, Stem Sample Capacity is 100uL
SDP
Catalog No. NC0623262
Encompass_Preferred
Coaxial Insert - 4mm TUBE W/50mm STEM. Includes one S-5-400-7 NMR tube to be used with the Coaxial Insert. 50mm Stem OD is 2mm, Stem Sample Capacity is 100?L Please note: we cannot guarantee the fit of any NMR tube other than the one that is provided with the insert.
Catalog No. NC0623262 Supplier Norell Inc Supplier No. NI5CCIB
Coaxial Insert - 4mm TUBE W/50mm STEM. Includes one S-5-400-7 NMR tube to be used with the Coaxial Insert. 50mm Stem OD is 2mm, Stem Sample Capacity is 100?L Please note: we cannot guarantee the fit of any NMR tube other than the one that is provided with the insert.