Learn More
World Precision Instrument Tinned Silver Alloy
SDP
Catalog No. 50822160
Encompass
Micro coaxial cables (MAXxxxx) are ideal for microelectrode fabrication and construction of similar research tools. The dual shielding eliminates electrical interference caused by radio frequencies (RF), electrostatic and microphonics (e.g., bending and vibration. Available with single or dual (twin) conductors.
Catalog No. 50-822-160 Supplier World Precision Instrument Supplier No. MAX3820
Add to Cart
Add to Cart
Micro coaxial cables (MAXxxxx) are ideal for microelectrode fabrication and construction of similar research tools. The dual shielding eliminates electrical interference caused by radio frequencies (RF), electrostatic and microphonics (e.g., bending and vibration. Available with single or dual (twin) conductors.