Learn More
Medchemexpress LLC Cob-187 | 1862177-86-3 | 95.0% | 302.41 | 25 MG
SDP
Catalog No. 5000395397
Encompass_Preferred
COB-187 is a potent, ATP-competitive, and selective inhibitor of GSK-3β. It inhibits GSK-3 through a reversible and Cysteine (Cys)-199-dependent mechanism. COB-187 also inhibits LPS-induced cytokine production and SARS-CoV-2 spike protein-induced CXCL10 production.
  • Potent, ATP-competitive and selective inhibitor of GSK-3β.
  • Inhibits GSK-3 through a reversible and Cysteine (Cys)-199-dependent mechanism.
  • Inhibits LPS-induced cytokine production.
  • Inhibits SARS-CoV-2 spike protein-induced CXCL10 production.
Catalog No. 50-003-95397 Supplier Medchemexpress LLC Supplier No. 25MGHY162675
COB-187 is a potent, ATP-competitive, and selective inhibitor of GSK-3β. It inhibits GSK-3 through a reversible and Cysteine (Cys)-199-dependent mechanism. COB-187 also inhibits LPS-induced cytokine production and SARS-CoV-2 spike protein-induced CXCL10 production.
  • Potent, ATP-competitive and selective inhibitor of GSK-3β.
  • Inhibits GSK-3 through a reversible and Cysteine (Cys)-199-dependent mechanism.
  • Inhibits LPS-induced cytokine production.
  • Inhibits SARS-CoV-2 spike protein-induced CXCL10 production.