Learn More
Strem, An Ascensus Company CAS# 136-52-7. 50g. Cobalt 2-ethylhexanoate in mineral spirits (12%Co). MFCD00072632
SDP
Supplier:  Strem, An Ascensus Company 27044950G
Encompass
CAS# 136-52-7. 50g. Cobalt 2-ethylhexanoate in mineral spirits (12%Co). MFCD00072632. Molecular Weight: 345.42. Molecular Formula: Co[OOCCH(C2H5)C4H9]2. Color/form: purple liq. Strem# 27-0449. http://www.strem.com/catalog/v/27-0449/
Catalog No. 50-901-11164