Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Cobalt

Catalog No. 266639500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-663-9500G 500 g
26-663-9100G 100 g
26-663-925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 26-663-9500G Supplier Sigma Aldrich Supplier No. 266639500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Cobalt powder has been used in the preparation of cobalt-based alloys and composites as anode materials. It can be also used:In the preparation of soft magnetic materials by powder metallurgical processes.As a catalyst in the transformation of ethanol to 1-butanol.

Specifications

CAS 7440-48-4
Boiling Point 2900°C
Density 8.9 g/mL at 25°C
Quantity 500 g
Molecular Formula Co
Linear Formula Co
MDL Number MFCD00010935
Molecular Weight (g/mol) 58.93 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.