Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Cobalt Carbonyl

Catalog No. 6081125G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-811-25G 25 g
60-811-5G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 60-811-25G Supplier Sigma Aldrich Supplier No. 6081125G
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Cobalt carbonyl [Co2(CO)8] is commonly used as a catalyst in the hydroformylation (oxo reaction) of alkenes.Along with pyridine, it can be used as a catalyst in the carboxylation of alkenes into corresponding acids and esters.It is employed as a key precursor in the preparation of cobalt platinum (CoPt3), cobalt sulfide (Co3S4) and cobalt selenide (CoSe2) nanocrystals.It is also used as a reagent in Pauson-Khand cyclizations and Nicholas reaction.

Specifications

CAS 10210-68-1
Molecular Formula C8Co2O8
Linear Formula Co2(CO)8
MDL Number MFCD00016024
Quantity 25 g
Synonym Cobalt tetracarbonyl dimer; Di-mu-carbonylhexacarbonyldicobal; Octacarbonyldicobalt; Dicobalt octacarbonyl
Molecular Weight (g/mol) 341.95 g/mol
Biological Activity Nervous System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.