Learn More
Strem, An Ascensus Company CAS# 10210-68-1. Cobalt carbonyl (Dicobalt octacarbonyl) (Stabilized with 1-5% hexanes). MFCD00016024. 100G
SDP
Catalog No. 5090111160
Encompass
CAS# 10210-68-1. 100G. Cobalt carbonyl (Dicobalt octacarbonyl) (Stabilized with 1-5% hexanes). MFCD00016024. Molecular Formula: C8Co2O8. Molecular Weight: 341.95. Color/Form: dark orange xtl. Stability: air sensitive, store cold (-18°C) Strem#: 27-0400 www.strem.com/catalog/v3/27-0400/
Catalog No. 50-901-11160 Supplier Strem, An Ascensus Company Supplier No. 270400100G
CAS# 10210-68-1. 100G. Cobalt carbonyl (Dicobalt octacarbonyl) (Stabilized with 1-5% hexanes). MFCD00016024. Molecular Formula: C8Co2O8. Molecular Weight: 341.95. Color/Form: dark orange xtl. Stability: air sensitive, store cold (-18°C) Strem#: 27-0400 www.strem.com/catalog/v3/27-0400/