Learn More
Strem, An Ascensus Company CAS# 10210-68-1. Cobalt carbonyl (Dicobalt octacarbonyl) (Stabilized with 1-5% hexanes). MFCD00016024. 25G
SDP
Supplier:  Strem, An Ascensus Company 27040025G
Encompass
CAS# 10210-68-1. 25G. Cobalt carbonyl (Dicobalt octacarbonyl) (Stabilized with 1-5% hexanes). MFCD00016024. Molecular Formula: C8Co2O8. Molecular Weight: 341.95. Color/Form: dark orange xtl. Stability: air sensitive, store cold (-18°C) Strem#: 27-0400 www.strem.com/catalog/v3/27-0400/
Catalog No. 50-901-11159