Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobalt Chloride Hexahydrate, >98%, (ACS Reagent Grade), MP Biomedicals

Catalog No. ICN19403580 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19403580 100 g
ICN19403525 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19403580 Supplier MP Biomedicals, Inc Supplier No. 0219403580
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7791-13-1 | Cl2CoH12O6 | 237.92 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7791-13-1
Molecular Formula Cl2CoH12O6
Molecular Weight (g/mol) 237.92
MDL Number MFCD00149652
InChI Key GFHNAMRJFCEERV-UHFFFAOYSA-L
IUPAC Name λ²-cobalt(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Co++]

Specifications

Physical Form Crystalline
Quantity 100 g
Assay Percent Range >98%
Formula Weight 237.9
Grade ACS Reagent
Chemical Name or Material Cobalt Chloride Hexahydrate
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.