Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobalt Chloride Hexahydrate, ACS Reagent, Reagents

Catalog No. 01338041
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-338-041 100 g
01-338-040 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-338-041 Supplier Reagents Holdings Llc Supplier No. C1181000100C2
Add to Cart
Add to Cart

Cobalt Chloride Hexahydrate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7791-13-1
Molecular Formula Cl2CoH12O6
Molecular Weight (g/mol) 237.92
MDL Number MFCD00149652
InChI Key GFHNAMRJFCEERV-UHFFFAOYSA-L
Synonym Cobalt(II) Chloride Hexahydrate, Cobaltous Chloride Hexahydrate
IUPAC Name λ²-cobalt(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Co++]

Specifications

Boiling Point 1049°C
Melting Point 56°C
Color Pink to Red
pH 4.9
Physical Form Solid
Quantity 100 g
Percent Purity 98%
Grade ACS
Packaging Glass Bottle
Specific Gravity 1.924
Chemical Name or Material Cobalt Chloride Hexahydrate, ACS Reagent
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.